4-(1-Pyrrolyl)benzonitril

Lieferant: Thermo Scientific

CC25516.250
MAYBCC25516.250EA 0 EUR
MAYBCC25516.250
4-(1-Pyrrolyl)benzonitril
4-(1-Pyrrolyl)benzonitril
Formel: C₁₁H₈N₂
MDL: MFCD00085164
CAS-Nummer: 23351-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR