4-Brom-2-thenoesäure

Lieferant: Thermo Scientific

Synonyme: 4-Bromo-2-thiophenecarboxylic acid

CC28501.250 CC28501DA
MAYBCC28501.250EA 0 EUR
MAYBCC28501.250 MAYBCC28501DA
4-Brom-2-thenoesäure
4-Brom-2-thenoesäure
Formel: C₅H₃BrO₂S
Molecular Weight: 207,05 g/mol
MDL: MFCD03422294
CAS-Nummer: 16694-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR