2-(4-Pyridinyl)-4-thiazolcarbonsäure
Lieferant: Thermo Scientific
CC34701DA
MAYBCC34701DAEA
0
EUR
MAYBCC34701DA
2-(4-Pyridinyl)-4-thiazolcarbonsäure
2-(4-Pyridinyl)-4-thiazolcarbonsäure
Formel:
C₉H₆N₂O₂S Molecular Weight: 206.22 g/mol |
MDL:
MFCD00171745 CAS-Nummer: 21278-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...