(1-Butyl)triphenylphosphoniumbromid
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
Butyltriphenylphosphonium bromide
CD00111.250
CD00111.1
CHMPCD00111.250EA
0
EUR
CHMPCD00111.250
MAYBCD00111.1
(1-Butyl)triphenylphosphoniumbromid
(1-Butyl)triphenylphosphoniumbromid
Formel:
H₃C(CH₂)₃P(C₆H₅)₃Br Molecular Weight: 399.31 g/mol Schmelzpunkt: 249…251 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011855 CAS-Nummer: 1779-51-7 EINECS: 217-219-4 UN: 3464 ADR: 6.1 III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...