5-(Chlormethyl)-3-(4-chlorphenyl)-1,2,4-oxadiazol

Lieferant: Thermo Scientific

CD02742CB CD02742DA
MAYBCD02742CBEA 0 EUR
MAYBCD02742CB MAYBCD02742DA
5-(Chlormethyl)-3-(4-chlorphenyl)-1,2,4-oxadiazol
5-(Chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole
Formel: C₉H₆Cl₂N₂O
MDL: MFCD00119077
CAS-Nummer: 57238-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR