α'-Chlor-α,α,α-trifluor-m-xylol

Lieferant: Thermo Scientific

CD03079.1 CD03079.10 CD03079.50
MAYBCD03079.1EA 0 EUR
MAYBCD03079.1 MAYBCD03079.10 MAYBCD03079.50
α'-Chlor-α,α,α-trifluor-m-xylol
α'-Chlor-α,α,α-trifluor-m-xylol
Formel: C₈H₆ClF₃
Siedepunkt: 95 °C (1013 hPa)
Dichte: 1,3079 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000908
CAS-Nummer: 705-29-3
EINECS: 211-884-4
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR