3-(2,6-Dichlorphenyl)-5-methyl-4-isoxazolcarbonsäureamid

Lieferant: Thermo Scientific

CD05279.50
MAYBCD05279.50EA 0 EUR
MAYBCD05279.50
3-(2,6-Dichlorphenyl)-5-methyl-4-isoxazolcarbonsäureamid
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxamide
Formel: C₁₁H₈Cl₂N₂O₂
MDL: MFCD00052211
CAS-Nummer: 23858-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR