1-Methyl-1-(2-nitro-4-(trifluormethyl)phenyl)hydrazin

Lieferant: Thermo Scientific

CD06991.50
MAYBCD06991.50EA 0 EUR
MAYBCD06991.50
1-Methyl-1-(2-nitro-4-(trifluormethyl)phenyl)hydrazin
1-Methyl-1-(2-nitro-4-(trifluoromethyl)phenyl)hydrazine
Formel: C₈H₈F₃N₃O₂
MDL: MFCD00052691
CAS-Nummer: 120940-43-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR