2,5-Dichlor-3-thiophensulfonylchlorid

Lieferant: Thermo Scientific

CD09480.0005
MAYBCD09480.0005EA 0 EUR
MAYBCD09480.0005
2,5-Dichlor-3-thiophensulfonylchlorid
2,5-Dichlor-3-thiophensulfonylchlorid
Formel: C₄HCl₃O₂S₂
MDL: MFCD00051665
CAS-Nummer: 56946-83-9
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR