4-Brom-2,5-dichlor-3-thiophensulfonylchlorid

Lieferant: Thermo Scientific

CD09555DA CD09555.10
MAYBCD09555DAEA 0 EUR
MAYBCD09555DA MAYBCD09555.10
4-Brom-2,5-dichlor-3-thiophensulfonylchlorid
4-Brom-2,5-dichlor-3-thiophensulfonylchlorid
Formel: C₄BrCl₃O₂S₂
MDL: MFCD00051664
CAS-Nummer: 166964-36-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR