4-(Trifluormethyl)phenylhydrazin
Lieferant: Thermo Scientific
CD11012.1
CD11012.10
MAYBCD11012.1EA
0
EUR
MAYBCD11012.1
MAYBCD11012.10
4-(Trifluormethyl)phenylhydrazin
4-(Trifluormethyl)phenylhydrazin
Formel:
C₇H₇F₃N₂ |
MDL:
MFCD00042508 CAS-Nummer: 368-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...