3-Chlor-2-fluorbenzamid

Lieferant: CHEMPUR

Synonyme: 3-Chlor-2-fluorbenzoesäureamid

CD11825.250 CD11825.1
CHMPCD11825.250EA 0 EUR
CHMPCD11825.250 MAYBCD11825.1
3-Chlor-2-fluorbenzamid
3-Chlor-2-fluorbenzamid
Formel: C₇H₅ClFNO
Molecular Weight: 173.57 g/mol
MDL: MFCD00042205
CAS-Nummer: 104326-94-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR