2,6-Dimethoxybenzaldehyd
Lieferant: Thermo Scientific
|
Achtung
|
CD11827.1
CD11827.10
MAYBCD11827.1EA
0
EUR
MAYBCD11827.1
MAYBCD11827.10
2,6-Dimethoxybenzaldehyd
2,6-Dimethoxybenzaldehyde
Formel:
C₉H₁₀O₃ Molecular Weight: 166.18 g/mol Siedepunkt: 285 Schmelzpunkt: 95…96 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00010862 CAS-Nummer: 3392-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...