4-(1,3-Oxazol-5-yl)anilin

Lieferant: Thermo Scientific

DFP00120.0100 DFP00120CB DFP00120DA
MAYBDFP00120.0100EA 0 EUR
MAYBDFP00120.0100 MAYBDFP00120CB MAYBDFP00120DA
4-(1,3-Oxazol-5-yl)anilin
4-(1,3-Oxazol-5-yl)anilin
Formel: C₉H₈N₂O
Molecular Weight: 160,1700 g/mol
MDL: MFCD00052192
CAS-Nummer: 1008-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR