N-(Cyanacetyl)morpholin

Lieferant: Thermo Scientific

Synonyme: 3-Morpholino-3-oxopropanenitrile, 3-(Morpholin-4-yl)-3-oxopropanenitrile

DP00099.0050
MAYBDP00099.0050EA 0 EUR
MAYBDP00099.0050
N-(Cyanacetyl)morpholin
N-(Cyanacetyl)morpholin
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154,17 g/mol
MDL: MFCD00023353
CAS-Nummer: 15029-32-0
EINECS: 239-112-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR