1-Chlor-4-iod-2,5-dimethoxybenzol

Lieferant: Thermo Scientific

Synonyme: 1-Chloro-2,5-dimethoxy-4-iodobenzene

DP00952.250
MAYBDP00952.250EA 0 EUR
MAYBDP00952.250
1-Chlor-4-iod-2,5-dimethoxybenzol
1-Chlor-4-iod-2,5-dimethoxybenzol
Formel: C₈H₈ClIO₂
CAS-Nummer: 90064-46-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR