3,5-Bis(trifluormethyl)benzoesäure
Lieferant: Thermo Scientific
DP01857.250
DP01857DA
MAYBDP01857.250EA
0
EUR
MAYBDP01857.250
MAYBDP01857DA
3,5-Bis(trifluormethyl)benzoesäure
3,5-Bis(trifluormethyl)benzoesäure
Formel:
C₉H₄F₆O₂ Molecular Weight: 258,12 g/mol Siedepunkt: 260 °C (1013 hPa) Schmelzpunkt: 140…144 °C Dichte: 1,42 g/cm³ (20 °C) |
MDL:
MFCD00000388 CAS-Nummer: 725-89-3 EINECS: 211-970-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...