Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat
Lieferant: Thermo Scientific
Synonyme:
Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
EN00291.1
EN00291EA
EN00291EE
MAYBEN00291.1EA
0
EUR
MAYBEN00291.1
MAYBEN00291EA
MAYBEN00291EE
Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat
Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat
Formel:
C₇H₁₁N₃O₂ |
MDL:
MFCD00051652 CAS-Nummer: 31037-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...