2-Amino-4-methylthiophen-3-carbonsäureamid

Lieferant: Thermo Scientific

GK00199.10
MAYBGK00199.10EA 0 EUR
MAYBGK00199.10
2-Amino-4-methylthiophen-3-carbonsäureamid
2-Amino-4-methylthiophene-3-carboxamide
Formel: C₆H₈N₂OS
Molecular Weight: 156,20 g/mol
MDL: MFCD00052594
CAS-Nummer: 4651-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR