3-Chlor-2-thenoesäure

Lieferant: Thermo Scientific

Synonyme: 3-Chlorthiophen-2-carbonsäure, 3-Chlor-2-thiophencarbonsäure

GK01306.1
MAYBGK01306.1EA 0 EUR
MAYBGK01306.1
3-Chlor-2-thenoesäure
3-Chlor-2-thenoesäure
Formel: C₅H₃ClO₂S
Molecular Weight: 162,6 g/mol
Schmelzpunkt: 187…188 °C
MDL: MFCD00043888
CAS-Nummer: 59337-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR