3-Chlor-2-thiophencarbonylchlorid

Lieferant: Thermo Scientific

GK01307 GK01307CB GK01307DA
MAYBGK01307EA 0 EUR
MAYBGK01307 MAYBGK01307CB MAYBGK01307DA
3-Chlor-2-thiophencarbonylchlorid
3-Chlor-2-thiophencarbonylchlorid
Formel: C₅H₂Cl₂OS
MDL: MFCD00068137
CAS-Nummer: 86427-02-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR