Methyl-4-brom-3-hydroxy-2-thiophencarboxylat

Lieferant: Thermo Scientific

Synonyme: Methyl 4-bromo-3-hydroxythiophene-2-carboxylate

GK01940.1 GK01940EA GK01940EE
MAYBGK01940.1EA 0 EUR
MAYBGK01940.1 MAYBGK01940EA MAYBGK01940EE
Methyl-4-brom-3-hydroxy-2-thiophencarboxylat
Methyl-4-brom-3-hydroxy-2-thiophencarboxylat
Formel: C₆H₅BrO₃S
Molecular Weight: 237.08 g/mol
MDL: MFCD00052081
CAS-Nummer: 95201-93-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR