3-Ethoxy-2-thenoesäure
Lieferant: Thermo Scientific
Synonyme:
3-Ethoxythiophene-2-carboxylic acid
GK02486.1
GK02486.10
MAYBGK02486.1EA
0
EUR
MAYBGK02486.1
MAYBGK02486.10
3-Ethoxy-2-thenoesäure
3-Ethoxy-2-thenoesäure
Formel:
C₇H₈O₃S Molecular Weight: 172.2 g/mol |
MDL:
MFCD00205205 CAS-Nummer: 139926-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...