Acetophenonoxim

Lieferant: CHEMPUR

Synonyme: Methyl phenyl ketoxime, 1-Phenyl-1-ethanone oxime, Acetophenone Oxime

GK03635.250 GK03635.1
CHMPGK03635.250EA 0 EUR
CHMPGK03635.250 MAYBGK03635.1
Acetophenonoxim
Acetophenonoxim
Formel: C₈H₉NO
Molecular Weight: 135.17 g/mol
MDL: MFCD00013931
CAS-Nummer: 613-91-2
EINECS: 210-360-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR