(2,4-Difluorphenyl)acetonitril

Lieferant: Thermo Scientific

Synonyme: 2,4-Difluorbenzylcyanid

HAN00045.1 HAN00045.10
MAYBHAN00045.1EA 0 EUR
MAYBHAN00045.1 MAYBHAN00045.10
(2,4-Difluorphenyl)acetonitril
(2,4-Difluorphenyl)acetonitril
Formel: C₈H₅F₂N
Molecular Weight: 153.13 g/mol
Siedepunkt: 90 °C (0.3 mmHg)
Dichte: 1.2503 g/cm³ (15 °C)
MDL: MFCD00009971
CAS-Nummer: 656-35-9
EINECS: 211-514-1
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR