4-Acetamido-3-chlorbenzolsulfonylchlorid

Lieferant: Thermo Scientific

HAN00134.250
MAYBHAN00134.250EA 0 EUR
MAYBHAN00134.250
4-Acetamido-3-chlorbenzolsulfonylchlorid
4-Acetamido-3-chlorbenzolsulfonylchlorid
Formel: C₈H₇Cl₂NO₃S
MDL: MFCD00052363
CAS-Nummer: 16761-18-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR