2-Piperazinonicotinonitril

Lieferant: Thermo Scientific

HTS04470.250 HTS04470DA
MAYBHTS04470.250EA 0 EUR
MAYBHTS04470.250 MAYBHTS04470DA
2-Piperazinonicotinonitril
2-Piperazinonicotinonitril
Formel: C₁₀H₁₂N₄
MDL: MFCD00190248
CAS-Nummer: 84951-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR