4-Chlor-8-(trifluormethyl)chinolin

Lieferant: CHEMPUR

JFD00637.250 JFD00637.1
CHMPJFD00637.250EA 0 EUR
CHMPJFD00637.250 MAYBJFD00637.1
4-Chlor-8-(trifluormethyl)chinolin
4-Chloro-8-(trifluoromethyl)quinoline
Formel: C₁₀H₅ClF₃N
MDL: MFCD00134578
CAS-Nummer: 23779-97-7
EINECS: 245-880-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR