1-Boc-piperidin-4-carbonsäure
Lieferant: Thermo Scientific
Synonyme:
1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid
JFD01115.250
JFD01115.1
MAYBJFD01115.250EA
0
EUR
MAYBJFD01115.250
MAYBJFD01115.1
1-Boc-piperidin-4-carbonsäure
1-Boc-piperidin-4-carbonsäure
Formel:
C₁₁H₁₉NO₄ |
MDL:
MFCD00076999 CAS-Nummer: 84358-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...