4,4'-Dimethylbiphenyl
Lieferant: CHEMPUR
Synonyme:
4,4'-Bitolyl, 1-Methyl-4-(4-methylphenyl)benzol
JFD01185.250
JFD01185.1
CHMPJFD01185.250EA
0
EUR
CHMPJFD01185.250
MAYBJFD01185.1
4,4'-Dimethylbiphenyl
4,4'-Dimethylbiphenyl
Formel:
C₁₄H₁₄ Molecular Weight: 182.27 g/mol Siedepunkt: 295 Schmelzpunkt: 117…122 °C Dichte: 1.102 g/cm³ (20 °C) |
MDL:
MFCD00008545 CAS-Nummer: 613-33-2 EINECS: 210-337-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...