2-Chlor-4-(methylsulfonyl)benzoesäure

Lieferant: Thermo Scientific

JFD01500.1 JFD01500EA JFD01500.50
MAYBJFD01500.1EA 0 EUR
MAYBJFD01500.1 MAYBJFD01500EA MAYBJFD01500.50
2-Chlor-4-(methylsulfonyl)benzoesäure
2-Chloro-4-(methylsulphonyl)benzoic acid
Formel: C₈H₇ClO₄S
MDL: MFCD00216496
CAS-Nummer: 53250-83-2
EINECS: 406-520-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR