4'-(Methylsulfonyl)acetophenon
Lieferant: Thermo Scientific
JFD01520.1
JFD01520EA
MAYBJFD01520.1EA
0
EUR
MAYBJFD01520.1
MAYBJFD01520EA
4'-(Methylsulfonyl)acetophenon
4'-(Methylsulfonyl)acetophenon
Formel:
C₉H₁₀O₃S |
MDL:
MFCD00025072 CAS-Nummer: 10297-73-1 EINECS: 233-672-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...