4-(Methylsulfonyl)phenol

Lieferant: CHEMPUR

JFD01524.250 JFD01524.1
CHMPJFD01524.250EA 0 EUR
CHMPJFD01524.250 MAYBJFD01524.1
4-(Methylsulfonyl)phenol
4-(Methylsulfonyl)phenol
Formel: C₇H₈O₃S
MDL: MFCD00047806
CAS-Nummer: 14763-60-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR