1-Brom-2,5-dichlor-3-fluorbenzol

Lieferant: CHEMPUR
Achtung

JFD01878.250 JFD01878.1
CHMPJFD01878.250EA 0 EUR
CHMPJFD01878.250 MAYBJFD01878.1
1-Brom-2,5-dichlor-3-fluorbenzol
1-Bromo-2,5-dichloro-3-fluorobenzene
Formel: C₆H₂BrCl₂F
Molecular Weight: 243.89 g/mol
MDL: MFCD00142585
CAS-Nummer: 202865-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR