(1,3-Dioxolan-2-ylmethyl)triphenylphosphoniumbromid

Lieferant: Thermo Scientific

Synonyme: ((1,3-Dioxolan-2-yl)methyl)triphenylphosphonium bromide

JFD03645.1
MAYBJFD03645.1EA 0 EUR
MAYBJFD03645.1
(1,3-Dioxolan-2-ylmethyl)triphenylphosphoniumbromid
(1,3-Dioxolan-2-ylmethyl)triphenylphosphoniumbromid
Formel: C₂₂H₂₂BrO₂P
MDL: MFCD00011966
CAS-Nummer: 52509-14-5
EINECS: 257-977-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR