1,3-Cyclopentandion
Lieferant: CHEMPUR
Synonyme:
Cyclopentane-1,3-dione
JFD03958.250
JFD03958.1
CHMPJFD03958.250EA
0
EUR
CHMPJFD03958.250
MAYBJFD03958.1
1,3-Cyclopentandion
1,3-Cyclopentandion
Formel:
C₅H₆O₂ Molecular Weight: 98,1 g/mol Schmelzpunkt: 149…152 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00001405 CAS-Nummer: 3859-41-4 EINECS: 223-372-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...