(4-Cyanbenzyl)triphenylphosphoniumbromid

Lieferant: Thermo Scientific

JFD04034.250 JFD04034.1
MAYBJFD04034.250EA 0 EUR
MAYBJFD04034.250 MAYBJFD04034.1
(4-Cyanbenzyl)triphenylphosphoniumbromid
(4-Cyanbenzyl)triphenylphosphoniumbromid
Formel: C₂₆H₂₁BrNP
CAS-Nummer: 26104-68-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR