3,5-Bis(trifluormethyl)benzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 3,5-Bis(trifluormethyl)benzolsulfochlorid, 3,5-Bis(trifluormethyl)benzol-1-sulfonylchlorid, 3,5-Bis(trifluormethyl)benzolsulfonsäurechlorid

JP00937DA JP00937.50
MAYBJP00937DAEA 0 EUR
MAYBJP00937DA MAYBJP00937.50
3,5-Bis(trifluormethyl)benzolsulfonylchlorid
3,5-Bis(trifluormethyl)benzolsulfonylchlorid
Formel: C₈H₃ClF₆O₂S
Schmelzpunkt: 36…38 °C
MDL: MFCD00014725
CAS-Nummer: 39234-86-1
EINECS: 254-371-0
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR