4,5-Dichlorimidazol

Lieferant: Thermo Scientific

Synonyme: 4,5-Dichlor-1H-imidazol

KM01364.1 KM01364.10 KM01364.50
MAYBKM01364.1EA 0 EUR
MAYBKM01364.1 MAYBKM01364.10 MAYBKM01364.50
4,5-Dichlorimidazol
4,5-Dichloroimidazole
Formel: C₃H₂Cl₂N₂
MDL: MFCD00005195
CAS-Nummer: 15965-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR