2-Brom-4,5-dichlorimidazol

Lieferant: Thermo Scientific

KM01467DA KM01467EA
MAYBKM01467DAEA 0 EUR
MAYBKM01467DA MAYBKM01467EA
2-Brom-4,5-dichlorimidazol
2-Bromo-4,5-dichloroimidazole
Formel: C₃HBrCl₂N₂
MDL: MFCD00067894
CAS-Nummer: 16076-27-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR