2-Fluorbenzolsulfonylchlorid
Lieferant: Thermo Scientific
KM01957.250
KM01957DA
MAYBKM01957.250EA
0
EUR
MAYBKM01957.250
MAYBKM01957DA
2-Fluorbenzolsulfonylchlorid
2-Fluorbenzolsulfonylchlorid
Formel:
C₆H₄ClFO₂S Siedepunkt: 242 °C (1013 hPa) Dichte: 1,47 g/cm³ (20 °C) |
MDL:
MFCD00042285 CAS-Nummer: 2905-21-7 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...