(1,2,2-Trimethylpropyliden)malononitril
Lieferant: Thermo Scientific
KM03717.1
KM03717.50
MAYBKM03717.1EA
0
EUR
MAYBKM03717.1
MAYBKM03717.50
(1,2,2-Trimethylpropyliden)malononitril
(1,2,2-Trimethylpropyliden)malononitril
Formel:
C₉H₁₂N₂ |
CAS-Nummer:
13017-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...