(1,2,2-Trimethylpropyliden)malononitril

Lieferant: Thermo Scientific

KM03717.1 KM03717.50
MAYBKM03717.1EA 0 EUR
MAYBKM03717.1 MAYBKM03717.50
(1,2,2-Trimethylpropyliden)malononitril
(1,2,2-Trimethylpropyliden)malononitril
Formel: C₉H₁₂N₂
CAS-Nummer: 13017-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR