2-Chlor-3-nitrothiophen

Lieferant: Thermo Scientific

KM05335.250 KM05335DA
MAYBKM05335.250EA 0 EUR
MAYBKM05335.250 MAYBKM05335DA
2-Chlor-3-nitrothiophen
2-Chloro-3-nitrothiophene
Formel: C₄H₂ClNO₂S
Molecular Weight: 163.58 g/mol
MDL: MFCD00052159
CAS-Nummer: 5330-98-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR