2-(Tributylstannyl)pyridin

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-(Tri-n-butylstannyl)pyridine

KM05480DA KM05480.10
MAYBKM05480DAEA 0 EUR
MAYBKM05480DA MAYBKM05480.10
2-(Tributylstannyl)pyridin
2-(Tributylstannyl)pyridin
Formel: C₁₇H₃₁NSn
Molecular Weight: 368,15 g/mol
MDL: MFCD00052052
CAS-Nummer: 17997-47-6
UN: 2788
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR