Thiazolidin-2-carbonsäure

Lieferant: Thermo Scientific

KM05539.50
MAYBKM05539.50EA 0 EUR
MAYBKM05539.50
Thiazolidin-2-carbonsäure
Thiazolidin-2-carbonsäure
Formel: C₄H₇NO₂S
MDL: MFCD00068005
CAS-Nummer: 65126-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR