3-(Tributylstannyl)pyridin

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-(Tri-n-butylstannyl)pyridine

KM05818.250 KM05818DA
MAYBKM05818.250EA 0 EUR
MAYBKM05818.250 MAYBKM05818DA
3-(Tributylstannyl)pyridin
3-(Tributylstannyl)pyridin
Formel: C₁₇H₃₁NSn
Molecular Weight: 368,15 g/mol
MDL: MFCD00052054
CAS-Nummer: 59020-10-9
UN: 2788
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR