2,1,3-Benzothiadiazol-4-ylamin

Lieferant: Thermo Scientific

Synonyme: 4-Aminobenzo-2,1,3-thiadiazole, 4-Aminopiazthiole, Benzo[c][1,2,5]thiadiazol-4-amine, 4-Amino-2,1,3-benzothiadiazole

KM06374.1
MAYBKM06374.1EA 0 EUR
MAYBKM06374.1
2,1,3-Benzothiadiazol-4-ylamin
2,1,3-Benzothiadiazol-4-ylamin
Formel: C₆H₅N₃S
Molecular Weight: 151,19 g/mol
MDL: MFCD00005810
CAS-Nummer: 767-64-6
EINECS: 212-186-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR