4-Brom-2-nitroanilin
Lieferant: Thermo Scientific
KM06414DA
KM06414.10
MAYBKM06414DAEA
0
EUR
MAYBKM06414DA
MAYBKM06414.10
4-Brom-2-nitroanilin
4-Brom-2-nitroanilin
Formel:
C₆H₅BrN₂O₂ Molecular Weight: 217,02 g/mol Siedepunkt: 309 °C (1013 hPa) Schmelzpunkt: 110…113 °C Dichte: 1,81 g/cm³ (20 °C) |
MDL:
MFCD00041312 CAS-Nummer: 875-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...