N-(2,4-Dichlorphenyl)-2-chloracetamid
Lieferant: Thermo Scientific
Synonyme:
2-Chloro-N-(2,4-dichlorophenyl)acetamide, N-(Chloroacetyl)-2,4-dichloroaniline
KM06507.250
KM06507.1
MAYBKM06507.250EA
0
EUR
MAYBKM06507.250
MAYBKM06507.1
N-(2,4-Dichlorphenyl)-2-chloracetamid
N-(2,4-Dichlorphenyl)-2-chloracetamid
Formel:
C₈H₆Cl₃NO Molecular Weight: 238.498 g/mol |
MDL:
MFCD00018888 CAS-Nummer: 6974-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...